Digital stopwatch, Traceable®
供应商: VWR Collection
VWR® Traceable®
VWRU62379-058EA 1755 CNY
VWRU62379-058
Digital stopwatch, Traceable®
定时器 数字秒表
Electronic digital stopwatch for precise timing and accurate laboratory results. Quartz-crystal precision is ideal for enzyme experiments, viscosity measurements, animal studies and all timed lab tests.
  • Timing capacity: 59 minutes, 59 seconds, 99 hundredths
  • Functions: Single action, time-out, continuous timing, and low battery indicator
  • Large LCD display is easy to read in all lighting conditions from total darkness to bright sunlight
  • Instrument is completely solid-state — no moving parts to wear out — no problems inherent in mechanical stopwatches
  • High-impact, plastic case engineered to fit comfortably in the hand or be worn with lanyard around the neck
Certifications: A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology. Calibrated by an ISO 17025 calibration laboratory accredited by A2LA.
Delivery information: Supplied with a 1,5 V silver oxide battery.