Combined fridge freezers
供应商: VWR Collection
VWR®
VWRU55703-512 VWRU55703-514 VWRU47747-226 VWRU47747-222
Combined fridge freezers
冰箱
VWR offers a complete line of specially designed and constructed explosion-proof and flammable material storage refrigerators, freezers, and combination units.
  • Feature a spark-proof, corrosion-resistant interior with steel encased electrical components
  • Adjustable temperature controls are mounted externally
  • Non-sparking magnetic door gaskets create a positive seal
Certifications: Meet OSHA and NFPA safety requirements