Precision weight sets, analytical
供应商: Troemner
TROE7233-1EA 21400 CNY
TROE7223-0 TROE7233-1 TROE7226-1W TROE7233-4 TROE7221-0W TROE7240-1T TROE7219-1W TROE7224-4 TROE7226-4W
Precision weight sets, analytical
称量产品 校准砝码
These weights are manufactured from the finest stainless steel including alloy 8 stainless steel. They are are designed with surface finish, markings, construction, material density, and magnetic properties. Weights of different classes are available.
Certifications: Accredited with National Voluntary Laboratory Accreditation Programme (NVLAP) under Lab Code 105013-0 and Statement of Accuracy.
Delivery information: The largest weight (500 g) and below are supplied in durable, high quality polycarbonate cases with a compartment for each weight within the case. There is also a compartment for a brush, forceps and a USB flash drive which contains the electronic certificate.