VWR® Talon®, Support stand, tripod base
供应商: VWR
VWRU12985-076EA 1341.07 CNY
VWRU12985-076
VWR® Talon®, Support stand, tripod base
支撑架
Tripod based support stand for use with VWR® Talon® lab frames.
  • Support stand base is constructed of cast iron with black enamel finish
  • Base features a built-in support rod holder with locking knob that accepts a 1,3 cm (¹/₂") Ø support rod (sold separately)