Microscope frosted slides, VWRⓇ
供应商: VWR, part of Avantor
VWR®
VWRU48312-004EA 177 CNY
VWRU48312-004 VWRU48382-173
Microscope frosted slides, VWRⓇ
载玻片和盖玻片 显微镜载片
High quality pre-cleaned sheet glass slides sandblasted on one end of one side, double frosted slides sandblasted on one end of both sides for marking specimens.