VWR®, Spatula, PTFE Coated
供应商: VWR
VWR®
VWRU82027-476EA 405.5 CNY
VWRU82027-476
VWR®, Spatula, PTFE Coated
样品勺
Stainless steel, PTFE coated, with wooden handle.
Sturdy, economical tools for handling, transferring, blending, or collecting chemical and biological samples. They can be used to transfer sticky or corrosive materials without scratching or damaging container surfaces.