VWR®Traceable® , Digital tachometer
供应商: VWR
VWRU23609-212EA 10385 CNY
VWRU23609-212
VWR®Traceable® , Digital tachometer
转速仪
High accuracy tachometer used to read speeds in centrifuges, motors and stirrers.
  • Five digit highly visible ³/₈"-LCD
  • Memory feature to recall the last reading
  • Laser accuracy anywhere from any rotating object
Certifications: ISO/IEC 17025:2005 (1750.01) accredited by A2LA indicating traceability to standards provided by NIST.
Delivery information: Supplied with Traceable® certificate, plastic case, adhesive tabs, internal light, four batteries and carrying case.