VWR® SpinPak®, Magnetic stirring bars
供应商: Avantor
VWR®
VWRU58948-047EA 479.53 CNY
VWRU58948-047
VWR® SpinPak®, Magnetic stirring bars
搅拌器 磁力搅拌子
These round stirring bars with a pivot ring are versatile and suitable for use with plastic containers. The Alnico magnet encapsulated in FDA grade PTFE is durable and inert. The removable pivot ring provides added versatility. It minimises the contact area of the bar to the vessel, reduces friction and lessens marring of plastic containers.
Delivery information: Set includes six bars in total, two each of 25,4×7,9 mm; 38,1×7,9 mm and 50,8×7,9 mm.