VWR® Traceable®, Hand tally counter
供应商: VWR
VWRU23609-102EA 431.32 CNY
VWRU23609-102
VWR® Traceable®, Hand tally counter
细胞计数器 人工细胞计数器
Tally counter is designed to count blood samples, take surveys, inventory items, total drops of liquid, receive lab samples and tally biomedical events.
  • Rugged, heavy-duty chrome-finished case with hinged finger loop for grip
  • Audible click that signals the count
  • Four figures, easy-to-use
Delivery information: Supplied with a loop.