VWR®, Autoclavable Bags, PP
供应商: VWR
VWR®
VWRU95042-548EA 1182.35 CNY
VWRU95042-548 VWRU95042-558 VWRU95042-556 VWRU95042-554 VWRU95042-552
VWR®, Autoclavable Bags, PP
废物处理袋
These sturdy, transparent 50 µm thick PP bags are suitable for autoclaving hazardous and nonhazardous materials and waste.
  • Highly resistant to tears and impact
  • Temperature resistant bags and autoclavable up to 135 °C
  • Twist ties allow proper closure for bag expansion and venting during sterilisation
Sturdy, temperature resistant bags, ideal for autoclaving hazardous, nonhazardous materials and waste. Supplied with twist ties that allow proper closure for bag expansion and venting during sterilisation.
Certifications: Meets strength requirements in ASTM D 1709-04 for 165 g dart drop test.
Delivery information: Supplied with twist ties.