VWR®, Bibulous Paper
供应商: VWR, part of Avantor
VWR®
VWRU28511-007EA 475 CNY
VWRU28511-007
VWR®, Bibulous Paper
纸张
High quality, highly absorbent paper, lint free, suitable for drying slides and small glass components.