Spatulas, micro, with colour-coded handles
供应商: VWR, part of Avantor
VWR®
VWRU57951-010EA 704 CNY
VWRU57951-010
Spatulas, micro, with colour-coded handles
样品勺
Small, flat-ended spatulas with stainless steel blade. Perfect for handling extremely small samples.
  • Spatulas have flat-ended blades
  • Four handy sizes in each pack
  • Handles are colour-coded in blue, white, green, and red for easy size identification
Use to dispense/transfer expensive chemicals, place particulate matter in weighing containers, apply micro amounts of adhesive for tacking down specimens, or mix small granular samples. Appropriate for use with balances, microscopes, or combustion analysers.
Delivery information: Pack of four includes four different blade widths: 0,1/0,3/0,5 and 1,0 mm.