VWR®Signature™, Incubators, reach-in
供应商: VWR
VWR®
VWRU35962-096EA 186020.82 CNY
VWRU35962-062 VWRU35962-096
VWR®Signature™, Incubators, reach-in
培养箱 标准恒温器
These incubators provide gentle air flow and minimises the space. And the chamber reinforced to support roller apparatus and shakers.
  • Microprocessor temperature control
  • Over temperature safety protection
  • Temperature uniformity to 0,8 °C
  • Prewired access panel for chart recorder
The air jacket design and heated outer door allows for uniform heat throughout the incubators. Both models incorporate a microprocessor controller to achieve precise temperature uniformity. An independent, secondary temperature controller offers the added security of over temperature protection. The chamber floors are reinforced to support roller apparatus or shakers.
Delivery information: Supplied with six shelves.