e-Core™ punch, Whatman™
供应商: Cytiva
WHATWB100052
e-Core™ punch, Whatman™
The e-Core™ punch is a compact, versatile, electric coring instrument designed for efficient sampling of Whatman™ FTA and FTA Elute™ matrices.
This semi-automated punch is designed to utilise the replaceable GE Healthcare Micro-Punch™ tips.
  • Razor-sharp cutting edge cleanly and precisely cores discs from FTA and FTA Elute, all but eliminating the creation of artifacts and potential cross-contamination between sampling
  • An ergonomic “joy stick” grip and simple operation provide comfort and efficiency, integrating smoothly into the laboratory sample throughput
  • The e-Core™ punch offers a combination of sampling features - coring, retrieving, and storing in a single operation, with decreased processing times
  • The e-Core™ ejection system allows the user to control the delivery of the ejected disc, and does not generate static, ensuring accurate disc delivery