Fiber glass roving, PYREX®
供应商: Corning Life Sciences
PYREX®
CORN3950EA 0 CNY
CORN3950
Fiber glass roving, PYREX®
玻璃棉
This fiber glass is made from fine glass fibers with diameters controlled to approximately 0,008 mm. This makes the glass pliable and easy to use. It contains minimal heavy metals and is suitable for use as a filtering medium.
Packaging: Supplied in the form of roving and packaged in 0,23 kg bags.