Anaesthesia induction chambers
供应商: VetEquip
VETQ941444EA 4698 CNY
VETQ941443 VETQ941444
Anaesthesia induction chambers
动物用品 动物麻醉系统
These chambers allow anaesthetic gases to be circulated throughout the chamber.
  • Durable, clear acrylic construction
  • Positive sealing gasket at lid to help prevent leakage
  • Large easy-to-use latches
  • Standard anaesthesia size inlet and outlet ports
  • Both ports on one side to reduce tubing in procedure area
  • Ports positioned above level where sleeping animals could create occlusions
  • Non-skid feet