VWR®Traceable®, Digital stopwatches, water-resistant / waterproof
供应商: VWR
VWR® Traceable®
VWRU61161-350EA 411.41 CNY
VWRU61161-350 VWRU62379-460
VWR®Traceable®, Digital stopwatches, water-resistant / waterproof
定时器 数字秒表
These stopwatches are perfect for lab and plant use. They are housed in a water-resistant/shock-resistant or waterproof/shockproof ABS plastic case. Water-resistant model is black. Waterproof model is yellow in colour.
  • Timing capacity: 23 hours, 59 minutes, 59 seconds (with a 1 second resolution)
  • Timing resolution is 0,01 seconds for the first 30 minutes
  • Functions: Single action, time-out, cumulative split, and continuous timing
  • Displays time-of-day in hours, minutes, and seconds, and features time-of-day alarm
  • Displays date and day of the week
  • Tactile-feel with 'click' switches provide positive action
Colour: Yellow or black
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Delivery information: Supplied with 1,5 V silver-oxide battery, lanyard and Traceable® certificate.