Affinity chromatography columns, StrepTrap™ HP
供应商: Cytiva
StrepTrap™
GEHE28-9075-46EA 0 CNY
GEHE28-9075-46 GEHE28-9075-48 GEHE29-0486-53 GEHE28-9075-47
Affinity chromatography columns, StrepTrap™ HP
色谱柱
StrepTrap™ HP is a ready to use HiTrap™ column prepacked with StrepTactin™ Sepharose™ High Performance for purifying Strep-tag II recombinant proteins.
  • Highly pure Strep-tag II recombinant proteins eluted in concentrated form and small volumes
  • Physiological conditions and mild elution preserves the activity of the target protein
  • Compatible with a wide range of reducing agents, detergents, denaturants and other additives
  • Easily regenerated with 0,5 M NaOH
Purification is performed under physiological conditions and the mild elution preserves the activity of the target protein.