Cell imaging dishes, with cover glass base
供应商: Eppendorf
EPPE0030740.009EA 1900 CNY
EPPE0030740.009 EPPE0030740.017
Cell imaging dishes, with cover glass base
培养皿 组织培养皿
Eppendorf cell imaging dishes ensure excellent results in high resolution microscopy of living and fixed cells. The bottom of the 35 mm dishes is made from cover glass. The central 18 mm cavity is lower than the level of the surrounding polystyrene dish, which enables the concentration of the cells during seeding on the glass surface and helps to reduce costs of antibodies and dyes. Plate handling and reference are easy, safe, and convenient due to a polygonal gripping zone and directional orientation marking.
  • TC treated coverglass surface for reliable results when working with adherent cells
  • Easy access to the complete imaging area with immersion lenses for alleviated handling and advanced compatibility with microscopes
  • 18 mm central cavity for defined growth and staining of cells for reduced costs of antibodies and dyes
  • Polygonal gripping zone for improved handling
  • Macroscopic orientation marks facilitate easy orientation
  • Low rim side walls allow for easy access for microinjection
  • Extraordinary planarity supports enhanced optical performance even during temperature shifts
Proof on non-cytotoxicity according to ISO 10993 specifications; lot-certified sterility according to ISO 11137 and 11135; sterility assurance level (SAL) of 10⁻⁶; lot-specific testing and certification for absence of detectable pyrogens, DNA, DNase and RNase; testing for cell attachment and cell growth with an anchorage-dependent cell line.