Anti-H2-K1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 25-D1.16]
供应商: BioLegend
BLEG141605EA 0 CNY
BLEG141605 BLEG141606
Anti-H2-K1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 25-D1.16]
抗体
Type: Primary
Antigen: H2-K1
Clonality: Monoclonal
Clone: 25-D1.16
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse