Anti-IGK Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RMK-45]

供应商: BioLegend
BLEG409506EA 0 CNY
BLEG409506
Anti-IGK Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RMK-45]
抗体
Type: Primary
Antigen: IGK
Clonality: Monoclonal
Clone: RMK-45
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Mouse
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR