Anti-IGHG1 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: HP6017]

供应商: BioLegend
BLEG409320EA 0 CNY
BLEG409320
Anti-IGHG1 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: HP6017]
抗体
Type: Primary
Antigen: IGHG1
Clonality: Monoclonal
Clone: HP6017
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Goat, Rabbit, Sheep, Horse
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR