罐,广口,带螺旋盖
供应商: VWR, part of Avantor
VWRI331-0112EA 194.4 CNY
VWRI331-0112 VWRI331-0187 VWRI331-0107 VWRI331-0109 VWRI331-0139 VWRI331-0103 VWRI331-0152 VWRI331-0108
罐,广口,带螺旋盖
油桶
HDPE, opaque with PP screw caps.
  • Space saving container, easy to fill
  • With tamper-evident screw caps
  • Excellent shock resistance
  • Temperature resistance: -10 to +80 °C (120 °C for short periods)
  • Can be supplied sterile
  • 紧凑型容器,便于填充
  • 带防盗螺旋盖
  • 卓越的防震性能
  • 温度耐受范围:- 10到+80°C(短时使用时,可耐受达120°C)
  • 可灭菌后交付
Ideal for water and general testing, filling and storage. Opaque, ensures reasonable visibility of the content level.