Anti-IgG2a Mouse Monoclonal Antibody [clone: AP1]

供应商: Santa Cruz
SANTSC-57761EA 0 CNY
SANTSC-57761
Anti-IgG2a Mouse Monoclonal Antibody [clone: AP1]
抗体
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: AP1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR