Test kits, COD, TNT plus®
供应商: Hach
HACHTNT820EA 971.1 CNY
HACHTNT820 HACHTNT821 HACHTNT82206 HACHTNT82306
Test kits, COD, TNT plus®
环境试验箱
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process.
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are colour coded by parameter and range
  • No reagent blank necessary