Automatic burettes, self-zeroing
供应商: Bel-Art
Scienceware®
BELAH379180112EA 4625 CNY
BELAH379180112
Automatic burettes, self-zeroing
滴定管
Self-zeroing glass burettes, Class B. Bottle, screw cap and round base made of PE.
  • Delivery end is fitted into a plastic push button stopcock which permits drop by drop delivery
  • Small screw clamp adjusts drop speed as needed for titration
  • PE filling tube extends from the top of the burette into the bottle reservoir
Burette have easy to read blue graduations against a white background. Complete with bottle and base.