Ammonia TNT plus kit
供应商: Hach
HACHTNT830EA 590 CNY
HACHTNT830
Ammonia TNT plus kit
环境试验箱
This kit is used for the determination of ammonia nitrogen by the salicylate method using TNTplus vials.
  • Reduce errors with bar coded vials and RFID
  • Innovative Vial
  • No reagent blank necessary