The BIOPURE system features a unique, award winning design incorporating a very high output water purification system with a large 350 liter storage reservoir. The system is designed to feed large or multiple automated endoscope reprocessors and washer disinfector installations with compliant purified water.
- Compact footprint to save space and facilitate easy installation
- Guaranteed bacterial specification, provided by onboard technology combined with easy system disinfection
- Reliable water quality – unique sanitisation design for purification of RO membranes as well as the local distribution loop
- Unique system operation protection using automated alarms and access to important control systems limited by 'E key'
Caution: Please note: Install in a properly ventilated area. All connections (Inlet, sanitization drain, reservoir drain, ringmain outlet and return) are 3/4” BSPP (female).