5-己腈 95%

Warning

ACRO259810050EA 0 CNY
ACRO259810050
5-己腈 95%
5-己腈
分子式: C₆H₉N
分子量: 95,15 g/mol
沸点: 162…163 °C (760 mmHg)
密度: 0,82 g/cm³
闪点: 40 °C
存储条件: Ambient
MDL编号: MFCD00067056
CAS号: 5048-19-1
UN号: 1993
ADR: 3,III

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4235 to 1.4285 (20°C, 589 nm)
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR