4-Epianhydrochlortetracycline hydrochloride, can be used as secondary standard

供应商: Thermo Fisher Scientific
ACRO268225000EA 0 CNY
ACRO268225000
4-Epianhydrochlortetracycline hydrochloride, can be used as secondary standard
标准品 Organic Standards
CAS No.: 158018-53-2

MDL No.: MFCD00143313
GHS Symbols: GHS07, GHS08,
Melting Point: 222 °C (D)
Appearance: Orange Powder
Storage Temperature: Ambient
Signal word: Warning
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR