Heptakis-6-bromo-6-deoxy-β-cyclodextrin

供应商: Thermo Scientific

ACRO299005000EA 0 CNY
ACRO299005000
Heptakis-6-bromo-6-deoxy-β-cyclodextrin
Heptakis-6-bromo-6-deoxy-beta-cyclodextrin

Appearance: Off-white Powder

分子式: C₄₂H₆₃Br₇O₂₈
溶点: 217…221 °C
存储条件: Ambient
MDL编号: MFCD00803289
CAS号: 53784-83-1

现在订购

产品技术规格

Appearance Off-white Powder
Melting point 217 to 221°C
NMR Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR