2-乙基吡咯 98%

供应商: Thermo Scientific
Warning

ACRO308770010EA 0 CNY
ACRO308770010
2-乙基吡咯 98%
2-乙基吡咯

Appearance: Clear yellow to brown Liquid

分子式: C₆H₉N
分子量: 95,15 g/mol
沸点: 142 °C (15 mmHg)
密度: 0,929 g/cm³
闪点: 62 °C
存储条件: Ambient
MDL编号: MFCD00060477
CAS号: 1551-06-0

现在订购

产品技术规格

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4985 to 1.5025 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR