Chloroform : isoamyl alcohol (24:1) (v / v) 用于分子生物学 DNase-, RNase-, protease-free
供应商: Thermo Scientific
|
Danger
|
ACRO327155000EA
0
CNY
ACRO327155000
Chloroform : isoamyl alcohol (24:1) (v / v) 用于分子生物学 DNase-, RNase-, protease-free
Chloroform : isoamyl alcohol
分子式:
CHCl₃,C₅H₁₂O 溶点: –63 °C 存储条件: Ambient |
MDL编号:
MFCD00171265 CAS号: 67-66-3 UN号: 1888 ADR: 6.1,III |
产品技术规格
Appearance | Clear solution |
GC |
(chloroform component) |
GC | ≥99.0 % (isoamyl alcohol component) |
Dnase,Rnase,Protease activity | none detected |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...