甲基三氧化铼(VII) 98%

Synonyms: 甲基铼(VII)三氧化物MTO

ACRO346961000EA 0 CNY
ACRO346961000 ACRO346960025 ACRO346965000
甲基三氧化铼(VII) 98%
甲基三氧化铼(VII)
分子式: CH₃O₃Re
分子量: 249,24 g/mol
溶点: 111 °C
存储条件: Ambient
MDL编号: MFCD00192332
CAS号: 70197-13-6

现在订购

产品技术规格

Appearance White to yellow to green to grey to blue-grey Powder or crystals
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR