Cresyl violet perchlorate 98%, 纯净
供应商: Thermo Scientific
|
Warning
|
ACRO405775000EA
0
CNY
ACRO405775000
Cresyl violet perchlorate 98%, 纯净
Cresyl violet perchlorate
Appearance: Green Powder
分子式:
C₁₆H₁₂ClN₃O₅ 分子量: 361,738 g/mol 溶点: 331…333 °C 存储条件: Ambient |
MDL编号:
MFCD00012671 CAS号: 41830-80-2 |
产品技术规格
Appearance | Green Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
UV Lambda max | 590 to 596 nm (in Methanol) |
Molar absorption coefficient | 67400 l/(mol.cm) (at Lambda max) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...