2-(Dimethylamino)ethyl acetate 98%

供应商: Thermo Scientific
Warning

ACRO408060250EA 0 CNY
ACRO408060250
2-(Dimethylamino)ethyl acetate 98%
2-(Dimethylamino)ethyl acetate

Appearance: Clear light yellow Liquid

分子式: C₆H₁₃NO₂
分子量: 131,18 g/mol
沸点: 152…154 °C
密度: 0,92 g/cm³
闪点: 47 °C
存储条件: Ambient
MDL编号: MFCD00041912
CAS号: 1421-89-2
UN号: 1992
ADR: 3,III

现在订购

产品技术规格

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4155 to 1.4175 (20°C, 589 nm)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR