N,N,N′,N′-四甲基二氨基氯磷酸 90% tech.

Danger

Synonyms: 双(二甲胺基)氯酸磷

ACRO172720100EA 0 CNY
ACRO172720100 ACRO172720500
N,N,N′,N′-四甲基二氨基氯磷酸 90% tech.
N,N,N′,N′-四甲基二氨基氯磷酸

Appearance: Clear colorless to light yellow Liquid

分子式: C₄H₁₂ClN₂OP
分子量: 170,58 g/mol
沸点: 246…247 °C (760 mmHg)
密度: 1,15 g/cm³
闪点: 119 °C
存储条件: Ambient
MDL编号: MFCD00008302
CAS号: 1605-65-8
UN号: 2922
ADR: 8,II

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4650 to 1.4680 (20°C, 589 nm)
Titration Argentometric 88 to 112 % (ex Cl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR