Basic Blue 24 (New Methylene Blue), 纯净

供应商: Thermo Scientific

ACRO192020100EA 0 CNY
ACRO192020100
Basic Blue 24 (New Methylene Blue), 纯净
Basic Blue 24 (New Methylene Blue)

Appearance: Dark brown to purple Crystalline powder

分子式: C₁₈H₂₂ClN₃S·0.5 ZnCl₂
分子量: 416,05 g/mol
存储条件: Ambient
MDL编号: MFCD00078135
CAS号: 6586-05-6

现在订购

产品技术规格

Appearance Dark brown to deep purple Crystalline powder
Infrared spectrum Conforms
Dye content ≥90 %
UV Lambda max 628 to 634 nm (c=5 mg/l, water)
UV Lambda max 588 to 594 nm (c=5 mg/l, water)
UV Lambda max 284 to 290 nm (c=5 mg/l, water)
UV Lambda max 242 to 248 nm (c=5 mg/l, water)
Molar extinction coefficient ≥40000 (c=5 mg/l, water) 628 to 634 nm
Molar extinction coefficient ≥36000 (c=5 mg/l, water) 588 to 594 nm
Molar extinction coefficient ≥28000 (c=5 mg/l, water) 284 to 290 nm
Molar extinction coefficient ≥16000 (c=5 mg/l, water) 242 to 248 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR