Dowex® 1X2 (Cl) 50-100 mesh 离子交换树脂

ACRO202951000EA 0 CNY
ACRO202951000 ACRO202955000
Dowex® 1X2 (Cl) 50-100 mesh 离子交换树脂
Dowex® 1X2 (Cl)
存储条件: Ambient
MDL编号: MFCD00132715
CAS号: 9085-42-1

现在订购

产品技术规格

Appearance Beads
Particle size 50 to 100 mesh
Particle size ≤10 % ( 180 µm)
Particle size ≤10 % ( 500 µm)
Wet volume capacity ≥0.7 meq/ml (Cl)
Water retention capacity 65 to 75 % (Cl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR