1,3,4-Triphenyl-4,5-dihydro-1H-1,2,4-triazol-5-ylidene

供应商: Thermo Scientific

ACRO363940010EA 0 CNY
ACRO363940010
1,3,4-Triphenyl-4,5-dihydro-1H-1,2,4-triazol-5-ylidene
1,3,4-Triphenyl-4,5-dihydro-1H-1,2,4-triazol-5-ylidene

Appearance: Beige Crystalline powder

分子式: C₂₀H₁₅N₃
溶点: 150 °C
存储条件: Freezer
MDL编号: MFCD00801868
CAS号: 166773-08-6

现在订购

产品技术规格

Appearance Beige Crystalline powder
Infrared spectrum Conforms
NMR Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR