O-(三甲基硅基)羟胺 97%

Danger

Synonyms: 氨氧基三甲基硅烷

ACRO367950050EA 0 CNY
ACRO367950050 ACRO367950010
O-(三甲基硅基)羟胺 97%
O-(三甲基硅基)羟胺

Appearance: Clear colorless Liquid

分子式: C₃H₁₁NOSi
分子量: 105,21 g/mol
沸点: 98…100 °C (760 mmHg)
密度: 0,86 g/cm³
闪点: 8 °C
存储条件: Ambient
MDL编号: MFCD00010872
CAS号: 22737-36-6
UN号: 1993
ADR: 3,II

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4060 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR