1,3-双(2,4,6-三甲基苯基)氯化咪唑氯盐 97%
|
Warning
|
Synonyms:
1,3-二(2,4,6-三甲苯)咪唑氯盐
ACRO378330010EA
0
CNY
ACRO378330050
ACRO378330010
1,3-双(2,4,6-三甲基苯基)氯化咪唑氯盐 97%
1,3-双(2,4,6-三甲基苯基)氯化咪唑氯盐
Appearance: Beige to pink-brown Powder
分子式:
C₂₁H₂₇ClN₂ 分子量: 342,91 g/mol 溶点: 280…286 °C 存储条件: Ambient |
MDL编号:
MFCD09039279 CAS号: 173035-10-4 |
产品技术规格
Appearance | Beige to pink-brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...