2-(Dicyclohexylphosphino)-2'-methylbiphenyl, 98%, (patented by MIT Cambridge U.S.A)

ACRO380080010EA 0 CNY
ACRO380080010
2-(Dicyclohexylphosphino)-2'-methylbiphenyl, 98%, (patented by MIT Cambridge U.S.A)
CAS No.: 251320-86-2

MDL No.: MFCD03094577
GHS Symbols: GHS07
Melting Point: 107°C - 111°C
Appearance: White Crystalline powder or crystals
Storage Temperature: Ambient
Signal word: Warning
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR