(S)-N-3-Cyanophenylalanine 95%, 98% ee

供应商: Thermo Scientific
Warning

ACRO362060010EA 0 CNY
ACRO362060010 ACRO362060050
(S)-N-3-Cyanophenylalanine 95%, 98% ee
(S)-N-3-Cyanophenylalanine

Appearance: White to light yellow Powder

分子式: C₁₀H₁₀N₂O₂
分子量: 190,202 g/mol
存储条件: Ambient
MDL编号: MFCD01860885
CAS号: 57213-48-6

现在订购

产品技术规格

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥94 %
Enantiomeric excess ≥97.5 % (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR