(S)-2-Isopropylsuccinic acid-1-methyl ester 95%, 98% ee

供应商: Thermo Scientific

ACRO362110010EA 0 CNY
ACRO362110010
(S)-2-Isopropylsuccinic acid-1-methyl ester 95%, 98% ee
(S)-2-Isopropylsuccinic acid-1-methyl ester

Appearance: Clear yellow Viscous liquid

分子式: C₈H₁₄O₄
存储条件: Refrigerator
CAS号: 208113-95-5

现在订购

产品技术规格

Appearance Clear yellow Viscous liquid
Infrared spectrum Conforms
GC ≥94 %
Enantiomeric excess ≥97.5 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR