Poly(caprolactone)triol, M.W. ∼ 300
供应商: Thermo Scientific
ACRO190730050EA
0
CNY
ACRO190730050
ACRO190730250
Poly(caprolactone)triol, M.W. ∼ 300
Poly(caprolactone)triol
分子式:
(C₆H₁₄O₃·C₆H₁₀O₂)x 沸点: 210 °C 溶点: –30 °C 密度: 1,07 g/cm³ 闪点: >199 °C 存储条件: Ambient |
MDL编号:
MFCD00084405 CAS号: 37625-56-2 |
产品技术规格
Appearance | Clear colorless Viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4750 to 1.4780 (20°C, 589 nm) |
Loss on drying | ≤2 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...