Vacuum pumps, Labsystem
供应商: Thermo Fisher Scientific
LABLVLP80-230EA 0 CNY
LABLVLP80-230 LABLVLP200-230
Vacuum pumps, Labsystem
真空转子蒸发器泵
These pumps achieves maximum solute recovery and designed to work with all SpeedVac™ concentrating and freeze drying systems and is applicable for vacuum concentrators.
  • Available for use with acids or DMSO/DMF
  • Standard oil drain kit simplifies oil change
  • Thermal overload switches with automatic reset
Three position gas ballast control optimizes pump performance to suit all applications. The large, sideview sight glass allows for convenient checking of proper oil levels at a glance.
Delivery information: Vacuum pump includes oil mist eliminator (OME190), filters (2) and line cord.