Titre plate shaker
供应商: Thermo Fisher Scientific
4625-1CEQ
BARN4625-1CEQEA 0 CNY
BARN4625-1CEQ
Titre plate shaker
振荡器 微孔板振荡器
These shakers provide shaking motion from gentle rotation to vigorous vortexing for ELISA, enzyme immunoassays, protein synthesis, pharmaceutical profiling among other applications.
  • Rugged, enameled steel shaker body with four nonskid rubber feet
  • Four platform finger slots for easy plate removal
  • Automatic shutoff
  • Vortexing motion thoroughly mixes each well
Independent start switch activates shaker for preset speed and time for repeatability in tests. Electronic timer can be set for continuous operation or timed shaking up to 5 min. Platform has 9 mm sides and holds from one to four 96-well standard or deep well plates.