Microtiter tubes
供应商: Porex
PORE845EA 0 CNY
PORE845
Microtiter tubes
管子 微管
For harvesting cells, cell uptakes and receptor binding.
Ideal for a wide variety of laboratory procedures, such as HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality
control, blood bank sample freezer storage, transport of specimens and reagents.