Concentric ring electrical steaming bath, precision
供应商: Thermo Scientific
BARN2897EA 0 CNY
BARN2897
Concentric ring electrical steaming bath, precision
浴锅 非搅拌水浴锅
This steaming bath is designed to gently warm samples, make concentrates, and melt solids such as agar.
  • Rugged epoxy-coated, corrosion-resistant stand
  • Copper-clad immersion heaters provide bath temperature up to 100 °C
  • Centre ring cover is 25 mm in diameter
  • Easy to operate with a four-position switch and three different heat settings
  • Adjustable water level regulator maintains bath depth at the desired level
Bath accommodates various sizes of glassware and evaporating dishes, including round-bottom flasks. Flange-formed stainless-steel rings nest smoothly and become progressively larger as each concentric ring is removed. Copper-clad immersion heaters provide bath temperatures up to 100 °C. Stainless-steel cabinets with flanged cabinet top for tabletop installations. Baths may also be flush-mounted in benchtop.
With support the height will be 330 mm. Requires 63 minutes for heating upto 100 °C.
订购须知 The bath requires hardwire installation by a qualified electrician and need to be hooked up to a continuous water source. Includes precision ring sets, constant water level device with 12,7 mm inlet for water supply, adjustable drain tube, and welded angle iron support frame with hot-rolled steel legs, fasteners and lock washers.